Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: ANSRTKLLW PKRESLLHEG LPKNHKAAKQ NVRGQEDNKA WKLKAVDAFK SAPLYQRPGY YSAPQTPLSP TPMFFPLEPS APSKLHSVTG IDTKDKSLKT VSSGAKKSFE LLSESDGALM EHPEVSQVRR KTVEFNLTDM PEIPENHLKE PLEQSPTNIH TTLKDHMDPY WALENRDEAH S (406-586 aa encoded by BC015220)
The retinal pigment epithelium (RPE) and choroid represent a differentiated system of the eye that sustains normal retinal health and function. Best vitelliform macular dystrophy, known as Best disease, is an early-onset autosomal dominant condition in which accumulation of lipofuscin-like material within and beneath the RPE leads to progressive loss of central vision. The lipofuscin-like material in the macular area appears as a yellow mass like the yolk of an egg that later becomes darker and irregular in color, a process known as "scrambling the egg". Best disease is frequently a reflection of mutations in the Bestrophin gene, which encodes a protein containing four putative transmembrane domains and localizes to the basolateral plasma membrane of RPE cells. Human Bestrophin forms oligomeric chloride channels that are sensitive to intracellular calcium. Missense mutations at the Bestrophin locus reduces or abolishes Bestrophin protein mediated membrane current. The human Bestrophin gene encodes a 585 amino acid protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BEST 1; Best disease; Best1V1Delta2; Bestrophin-1; Bestrophin1; TU15B; Vitelliform macular dystrophy protein 2; VMD 2
Gene Aliases: ARB; BEST; BEST1; BMD; RP50; TU15B; VMD2
UniProt ID: (Human) O76090
Entrez Gene ID: (Human) 7439
Molecular Function:
ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support