Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat and Bovine
Clathrin-mediated endocytosis is the pathway by which many receptors for nutrients and hormones are internalized to be recycled or down-regulated. During formation of clathrin coated membranes, clathrin co-assembles with heterotetrameric molecules known as assembly polypeptides (APs) or adaptors which form a layer of protein coat between the clathrin lattice and the membrane. There are two characterized adaptors AP1 and AP2. AP1 is associated with clathrin coated vesicles at the trans-Golgi network and AP2 is associated with the endocytic clathrin coated vesicles at the plasma membrane and has been shown to specifically interact with Shc and EGF receptor. AP2 is composed of four subunits, two separate ~100 kDa gene products with similar domain structures (alpha and beta adaptin) and a ~50 and ~17 kDa subunit. There are two alpha-adaptin genes, alpha A and alpha C which have a tissue specific pattern of expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adapter-related protein complex 2 beta subunit; adapter-related protein complex 2 subunit beta; adaptin, beta 2 (beta); Adaptor protein complex AP-2 subunit beta; adaptor related protein complex 2, beta 1 subunit; Adaptor-related protein complex 2 subunit beta; adaptor-related protein complex 2, beta 1 subunit; AP-2 complex subunit beta; AP105B; Beta-2-adaptin; Beta-adaptin; beta2-adaptin; Clathrin assembly protein complex 2 beta large chain; clathrin-associated/assembly/adaptor; clathrin-associated/assembly/adaptor protein, large, beta 1; Plasma membrane adaptor HA2/AP2 adaptin beta subunit; testicular tissue protein Li 22
Gene Aliases: 1300012O03Rik; ADTB2; AI788979; AP105B; AP2-BETA; AP2B1; CLAPB1
UniProt ID: (Human) P63010, (Mouse) Q9DBG3
Entrez Gene ID: (Human) 163, (Mouse) 71770
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support