Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Specificity: This antibody binds to Bet v 1.
May be a general steroid carrier protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: allergen Bet v 1-A; Allergen Bet v I-A; BETVI; Major pollen allergen Bet v 1-A
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support