Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Purity is > 95% by SDS-PAGE.
Bif1 gene encodes a protein similar to a protein in rodents which is induced by bone morphogenic protein 2 in vitro and may be involved in BMP2-induced transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bax-interacting factor 1; Bif-1; BIF1; endophilin b1; Endophilin-B1; protein phosphatase 1, regulatory subunit 70; SH3 domain-containing GRB2-like protein B1; SH3-containing protein SH3GLB1; SH3-domain GRB2 like endophilin B1; SH3-domain GRB2-like endophilin B1; testicular tissue protein Li 172
Gene Aliases: Bif-1; CGI-61; dJ612B15.2; KIAA0491; PPP1R70; SH3GLB1
UniProt ID: (Human) Q9Y371
Entrez Gene ID: (Human) 51100
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support