Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
BGN is a small cellular or pericellular matrix proteoglycan that is closely related in structure to two other small proteoglycans, decorin and fibromodulin. The protein and decorin are thought to be the result of a gene duplication. Decorin contains one attached glycosaminoglycan chain, while this protein probably contains two chains. For this reason, this protein is called biglycan. This protein is thought to function in connective tissue metabolism by binding to collagen fibrils and transferring growth factor-beta. It may promote neuronal survival.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Biglycan; bone/cartilage proteclycan 1; Bone/cartilage proteoglycan I; bone/cartilage proteoglycan-I; dermatan sulphate proteoglycan I; PG S1; PG-S1; small leucine-rich protein 1A; Small proteoglycan I (biglycan), bone (BSPG1) (bone/cartilage proteclycan 1 precursor)
Gene Aliases: BG; BGN; BSPG1; DSPG1; PG-S1; PGI; SLRR1A
UniProt ID: (Human) P21810, (Mouse) P28653, (Rat) P47853
Entrez Gene ID: (Rabbit) 100008663, (Human) 633, (Mouse) 12111, (Rat) 25181
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support