Invitrogen
FIGURE: 1 / 1
The PA1-73007 antibody reacts with Borrelia burgdorferi.
PA1-73007 has been successfully used in Immunofluorescence applications.
The PA1-73007 antibody was raised against B. burgdorferi whole cell preparation.
Borrelia burgdorferi is a spirochete and the cause of Lyme disease, a tick transmitted illness of humans and animals. B. burgdorferi may persist in humans and animals for months or years following initial infection, despite a robust humoral immune response. B. burgdorferi resembles other spirochetes in that it is a highly specialized, motile, two-membrane, spiral shaped bacteria which lives primarily as an extracellular pathogen. B. burgdorferi has an unusual genome compared with other eubacteria which includes a linear chromosome approximately one megabase in size and numerous linear and circular plasmids.
Protein Aliases: B. burgdorferi; Borrelia; Lyme disease