Invitrogen
The MA1-7284 antibody reacts with Borrelia burgdorferi Osp B in bacterial and infected tissue samples.
MA1-7284 has been successfully used in ELISA and Immunofluorescence applications.
The MA1-7284 antibody was raised against RV Osp B antigen.
Borrelia burgdorferi is a spirochete and the cause of Lyme disease, a tick transmitted illness of humans and animals. B. burgdorferi may persist in humans and animals for months or years following initial infection, despite a robust humoral immune response. B. burgdorferi resembles other spirochetes in that it is a highly specialized, motile, two-membrane, spiral shaped bacteria which lives primarily as an extracellular pathogen. B. burgdorferi has an unusual genome compared with other eubacteria which includes a linear chromosome approximately one megabase in size and numerous linear and circular plasmids.
Protein Aliases: B. burgdorferi; Borrelia; Lyme disease