Invitrogen
The MA1-7407 antibody reacts with Bovine Papillomavirus in viral and infected tissue samples.
MA1-7407 has been successfully used in ELISA applications.
The MA1-7407 antibody was raised against a BPV preparation.
Papillomaviruses are a diverse group of DNA-based viruses that infect the skin and mucous membranes of humans and a variety of animals. 100 different HPV types have been identified to date. Various types of HPV may cause warts while others may cause a subclinical infection resulting in precancerous lesions. All HPVs are transmitted by skin-to-skin contact.
Protein Aliases: BPV