Invitrogen
FIGURE: 1 / 2
This antibody reacts with BPV type 1 E2 protein and has been mapped to amino acids 310-410.
This is a conformational antibody and does not work in Western blot analysis or under other denaturating conditions.
Papillomaviruses are a diverse group of DNA-based viruses that infect the skin and mucous membranes of humans and a variety of animals. 100 different HPV types have been identified to date. Various types of HPV may cause warts while others may cause a subclinical infection resulting in precancerous lesions. All HPVs are transmitted by skin-to-skin contact.
Protein Aliases: BPV; BPV Protein E2; BPV-1; BPV1-E2