Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KLPNLKFLDA QKVTRQEREE ALVRGVFMKV VKPKASSEDV ASSPERHYTP LPSASRELTS HQGVLGKCRY VYYGKNSEGN RFIRDDQ
Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 25%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Leucine-rich melanocyte differentiation-associated protein
Gene Aliases: C10orf11; CDA017; LRMDA
UniProt ID: (Human) Q9H2I8
Entrez Gene ID: (Human) 83938
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support