Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: Target recombinant protein.
Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (88%), Chicken (89%), Rhesus Monkey (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Cell cycle associated protein capable of promoting cell proliferation through the activation of CDK2 at the G1/S phase transition.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cdk-associated cullin 1; Cdk-Associated Cullin1; CDK2-associated and cullin domain-containing protein 1; CDK2-associated, cullin domain 1; cullin
Gene Aliases: C10orf46; CAC1; CACUL1; RGD1308127
UniProt ID: (Human) Q86Y37, (Rat) Q5XI53
Entrez Gene ID: (Human) 143384, (Rat) 365493
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support