Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: RAIAYQHSGHL ESKDINQEEL RALEEVEMKL QKNFLTQREN TIAGANHTHT FYGHSHHSHH GHPSHQSHSL PNRRH
Highest antigen sequence identity to the following orthologs - mouse 49%, rat 28%.
C10orf62 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Uncharacterized protein C10orf62
Gene Aliases: bA548K23.1; C10orf62
UniProt ID: (Human) Q5T681
Entrez Gene ID: (Human) 414157
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support