Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MDPTCSSECI YNLIPSDLKE PPQPPRYISI FKATVKDDMQ KAKTAMKTMG PAKVEVPSPK DFLKKHSKEK TLPPKKNFDR NVPKKPAVPL KTDHPVMGIQ SGKNFINTNA ADIIMGVAKK PKPIYVDKRT GDKHDLEPSG LVPKYINKKD YGVTPEYICK RNEEIKKAQE DYDRYIQENL KKAAMKRLSD EEREAVLQGL KKNWEEVHKE FQSLSVFIDS IPKKIRKQRL EEEMKQLEHD IGIIEKHKII YIANNA
Enkurin interacts with transient receptor potential canonical.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Enkurin
Gene Aliases: C10orf63; CFAP106; ENKUR
UniProt ID: (Human) Q8TC29
Entrez Gene ID: (Human) 219670
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support