Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase PAAT; PAAT; Protein associated with ABC transporters
Gene Aliases: C10orf88; PAAT
UniProt ID: (Human) Q9H8K7
Entrez Gene ID: (Human) 80007
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support