Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
LINC02870 is a novel lncRNA, whose role in tumors has not been reported. LINC02870 was determined to be a possible oncogene in HCC. Upregulated expressions of LINC02870 were found in our human hepatocellular carcinoma (HCC) samples and overexpression of LINC02870 promoted the growth, migration, and invasion of HCC cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C10orf91 Uncharacterized protein
Gene Aliases: bA432J24.4
Entrez Gene ID: (Human) 170393
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support