Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSELTKELME LVWGTKSSPG LSDTIFCRWT QGFVFSESEG SALEQFEGGP CAVIAPVQAF LLKKLLFSSE KSSWRDCSEE EQKELLCHTL CDILESACCD HSGSYCLVSW LRGKTTEETA SISGSPAESS CQVEHSSALA VEELGFERFH ALIQKRSFRS LPELKDAVLD QYSMWGNKFG VLLFLYSVLL TKGIENIKNE IEDASEPLID PVYGHGSQSL INLLLTGHAV SNVWDGDREC SGMKLLGIHE QAAVGFLTLM EALRYCKVGS YLKSPKFPIW IVGSETHLTV FFAKDMALVA PEAPSEQARR VFQTYDPEDN GFIPDSLLED VMKALDLVSD PEYINLMKNK LDPEGLGIIL LGPFLQEFFP DQGSSGPESF TVYHYNGLKQ SNYNEKVMYV EGTAVVMGFE DPMLQTDDTP IKRCLQTKWP YIELLWTTDR SPSLN
The protein encoded by this gene contains a caspase-associated recruitment domain and may function in apoptosis. It has been identified as a tumor suppressor in lung and gastric cancers, and a polymorphism in the gene may be associated with gastric cancer risk. Alternative splicing of this gene results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CARD-containing protein; caspase recruitment domain containing pro-apoptotic protein; Dermal papilla-derived protein 5; Deubiquitinating enzyme MINDY-3; family with sequence similarity 188, member A; Protein CARP; RP11-394I23.1; Ubiquitin carboxyl-terminal hydrolase MINDY-3
Gene Aliases: C10orf97; CARP; DERP5; FAM188A; MINDY3; MST126; MSTP126; My042
UniProt ID: (Human) Q9H8M7
Entrez Gene ID: (Human) 80013
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support