Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: QERYDLRNIV QPKPLPSQFG HYFETTYDTS YNNKMPLSTH RFKREPHWFP GHQPELDPPR YKCTEKSTYM NSYSKP
Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 87%.
C11orf1 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cilia- and flagella-associated protein 68; UPF0686 protein
Gene Aliases: C11orf1; CFAP68
UniProt ID: (Human) Q9H5F2
Entrez Gene ID: (Human) 64776
Molecular Function:
calmodulin-related
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support