Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSRIPLGKVL LRNVIRHTDA HNKIQEESDM WKIRELEKQM EDAYRGTKRK MLPSSSSRMR SDGFDEESQR YYWRPKNEIS
Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
C11orf57 has three isoforms with MW 30-34 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NKAP domain containing protein 1; Uncharacterized protein NKAPD1
Gene Aliases: C11orf57; NKAPD1
UniProt ID: (Human) Q6ZUT1
Entrez Gene ID: (Human) 55216
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support