Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
C11orf65 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial fission factor interactor; Protein MFI
Gene Aliases: C11orf65; MFI
UniProt ID: (Human) Q8NCR3
Entrez Gene ID: (Human) 160140
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support