Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSAHMSGLEI MDEDQLIKDV LDKFLNCHEQ TYDEEFLNTF THLSQEDHVS KRGVFGTDSS E
Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 57%.
The exact function of C11orf74 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Intraflagellar transport-associated protein; Protein HEPIS
Gene Aliases: C11orf74; HEPIS; IFTAP; NWC
UniProt ID: (Human) Q86VG3
Entrez Gene ID: (Human) 119710
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support