Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RYALTTTLSA RVARLNPTWN HPDQDTEAGF KRAMDLVQEE FLQRLDFYQH SWLPARALVE EALAQRFQVD P
Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.
C12orf10 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: melanocyte proliferating gene 1; melanocyte related; MYG1 exonuclease
Gene Aliases: C12orf10; Gamm1; MST024; MSTP024; MYG; MYG1
UniProt ID: (Human) Q9HB07
Entrez Gene ID: (Human) 60314
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support