Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EKVYHSLTYL GQKLGGQSFF SRKDSIRTIY TSLHNELKKV VTGRGALGGT APHVEELLSH LSEQLCFFVQ ARMEMADFYE KMYTLSTQKF INA
As part of the KICSTOR complex functions in the amino acid-sensing branch of the TORC1 signaling pathway. Recruits, in an amino acid-independent manner, the GATOR1 complex to the lysosomal membranes and allows its interaction with GATOR2 and the RAG GTPases. Functions upstream of the RAG GTPases and is required to negatively regulate mTORC1 signaling in absence of amino acids. In absence of the KICSTOR complex mTORC1 is constitutively localized to the lysosome and activated. The KICSTOR complex is also probably involved in the regulation of mTORC1 by glucose.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KICSTOR complex protein C12orf66; KICSTOR subunit 2; UPF0536 protein
Gene Aliases: C12orf66; KICS2
UniProt ID: (Human) Q96MD2
Entrez Gene ID: (Human) 144577
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support