Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: 293T, Mouse kidney, Mouse heart.
Immunogen sequence: ADFASPFHER HFHYEEHLER MKRRSSASVS DSSGFSDSES ADSLYRNSFS FSDEKLNSPT D
Modulates the activity of cell cycle-specific kinases. Enhances CDK1 activity. May contribute to the regulation of the cell cycle. May inhibit growth of glioma cells by promoting arrest of mitotic progression at the G2/M transition. Fibrogenic factor contributing to the pathogenesis of renal fibrosis through fibroblast activation.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Regulator of cell cycle RGCC; Response gene to complement 32 protein; RGC-32
Gene Aliases: 1190002H23Rik; bA157L14.2; C13orf15; RGC-32; RGC32; RGCC
UniProt ID: (Human) Q9H4X1, (Mouse) Q9DBX1, (Rat) Q9Z2P4
Entrez Gene ID: (Human) 28984, (Mouse) 66214, (Rat) 117183
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support