Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Human: 100%
SPACA7 is a protein coding gene involved in fertilization. Seems not to play a direct role in sperm-egg binding or gamete fusion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: protein SPACA7; Sperm acrosome-associated protein 7
Gene Aliases: C13orf28; SPACA7
UniProt ID: (Human) Q96KW9
Entrez Gene ID: (Human) 122258
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support