Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKTLFEEIK ASIKNNYNQD RSFCRPVLPW GGVFTIKAGR KAVSCTPLYV EIRLKNTCTI DGFLMLLYVI LNENENFPRE LSLHFGREFV DCFLYLMDTY SFTTVKLLWI WDKMEKQQYK SEVHKASLII DLFGNEHDNF TKNLENLMST IQESYCSNWR CPTRVQEDQQ RTININPPQE IPHGNLIRLA VNELFCSKIE LCEEHGCGGL REFSQRIFCH GAPPFVVLNM QHWKSEDLAY VPYYLDLSDH KYLLEGATLF NKEEHHYSAA FQIGGHWMHY DGLRNVNLIL LNKPPEFLLL SSLVYIRATE K (1-310 aa encoded by BC105030)
C14orf28 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dopamine receptor interacting protein 1; Dopamine receptor-interacting protein 1; Uncharacterized protein C14orf28
Gene Aliases: c14_5270; C14orf28; DRIP-1; DRIP1
UniProt ID: (Human) Q4W4Y0
Entrez Gene ID: (Human) 122525
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support