Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RCCCCACKS ETNGGNTGSQ GGNPPPSTPI TVTGHGLAVQ SSEQLLHVIY QRVDKAVGLA EAALGLARAN NELLKRLQEE VGDLRQGKVS IPDEDGESRA HSSPPEEPGP LKESPGEAFK ALSAVEEECD SVGSGVQVVI EELRQLGAAS VGPGPLGFPA TQRDMRLPGC TLAASEAAPL LNPLVDDYVA SEGAVQRVLV PAYAKQLSPA TQLAIQRATP ETGPENGTKL PPPRPEDMLN AAAALDSALE ESGPGSTGEL RHSLGLTVSP CRTRGSGQKN SRRKRDLVLS KLVHNVHNHI TNDKRFNGSE SIKSSWNISV VKFLLEKLKQ ELVTSPHNYT DKELKGACVA YFL (18-369 aa encoded by BC014299)
C14orf93 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Uncharacterized protein C14orf93
Gene Aliases: C14orf93
UniProt ID: (Human) Q9H972
Entrez Gene ID: (Human) 60686
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support