Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MASGDFCSP GEGMEILQQV CSKQLPPCNL SKEDLLQNPY FSKLLLNLSQ HVDESGLSLT LAKEQAQAWK EVRLHKTTWL RSEILHRVIQ ELLVDYYVKI QDTNVTSEDK KFHETLEQRL LVTELMRLLG PSQEREIPPL LGLEKADLLE LMPLSEDFVW MRARLQQEVE EQLKKKCFTL LCYYDPNSDA DSETVKAAKV WKLAEVLVGE QQQCQDAKSQ QKEQMLLLEK KSAAYSQVLL RCLTLLQRLL QEHRLKTQSE LDRINAQYLE VKCGAMILKL RMEELKILSD TYTVEKVEVH RLIRDRLEGA IHLQEQDMEN SRQVLNSYEV LGEEFDRLVK EYTVLKQATE NKRWALQEFS KVYR (1-363 aa encoded by BC001916)
This gene encodes a subunit of the centrosome complex termed the human augmin complex. The encoded protein localizes to the spindle microtubules and may play a role in mitotic spindle assembly and maintenance of centrosome integrity during cell division. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HAUS augmin-like complex subunit 4; HAUS augmin-like complex, subunit 4; homologous to Augmin subunits
Gene Aliases: 9430093H08Rik; AU016300; C14orf94; D14Ertd500e; HAUS4
UniProt ID: (Human) Q9H6D7, (Mouse) Q8BFT2
Entrez Gene ID: (Human) 54930, (Mouse) 219072
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support