Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a subunit of the centrosome complex termed the human augmin complex. The encoded protein localizes to the spindle microtubules and may play a role in mitotic spindle assembly and maintenance of centrosome integrity during cell division. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HAUS augmin-like complex subunit 4; HAUS augmin-like complex, subunit 4; homologous to Augmin subunits
Gene Aliases: 9430093H08Rik; AU016300; C14orf94; D14Ertd500e; HAUS4; RGD1305288
UniProt ID: (Human) Q9H6D7, (Mouse) Q8BFT2
Entrez Gene ID: (Human) 54930, (Mouse) 219072, (Rat) 305882
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support