Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: NRKNTKTSPR GEGTAPPFSA RPCVWTLCEM LSILALVGVL HPFYRSNNQV
Sequence homology: Human: 100%
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Entrez Gene ID: (Human) 145858
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support