Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 93%; Dog: 79%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rat: 100%
Component of the protein machinery at the inhibitory synapses, probably acting as a scaffold. Inhibitory synapses dampen neuronal activity through postsynaptic hyperpolarization. This synaptic inhibition is fundamental for the functioning of the central nervous system, shaping and orchestrating the flow of information through neuronal networks to generate a precise neural code.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Inhibitory synaptic factor 1; InSyn1
Gene Aliases: C15orf59; INSYN1
UniProt ID: (Human) Q2T9L4
Entrez Gene ID: (Human) 388135
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support