Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EMADFLRIK LKPLDKVTKS PASSRAEKKA EPPPSKPTVA KTGLALIKDC CGATQCNIM (147-204 aa encoded by BC008967)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bMERB domain-containing protein 1; migration inhibitory protein; Uncharacterized protein C16orf45 homolog
Gene Aliases: BMERB1; C16orf45; MINP
UniProt ID: (Human) Q96MC5, (Rat) Q5FVJ5
Entrez Gene ID: (Human) 89927, (Rat) 360459
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support