Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product # 26086-1-AP is a rabbit polyclonal antibody raised against the full-length of human C16orf61. The MW of this protein is 9 kDa, and this antibody specifically recognizes the 9 kDa protein.
Immunogen sequence: MHPDLSPHL HTEECNVLIN LLKECHKNHN ILKFFGYCND VDRELRKCLK NEYVENRTKS REHGIAMRKK LFNPPEESEK (1-79 aa encoded by BC032631 )
The function of C16orf61 remains largely unknown. It may be involved in cytochrome c oxidase biogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-x(9)-C motif containing 2; COX assembly mitochondrial protein 2 homolog
Gene Aliases: 2310061C15Rik; C16orf61; CMC2; DC13
UniProt ID: (Human) Q9NRP2
Entrez Gene ID: (Human) 56942
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support