Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEERKEEGE AEIQEHGPEH WFSKWERQCL AEAEQDEQLP PELQEEAAAA AQPEHKQQKL WHLFQNSATA VAQLYKDRVC QQPGLSLWVP FQNAATAVTN LYKESVDTHQ RSFDIGIQIG YQRRNKDVLA WVKKRRRTIR REDLISFLCG KVPPPRNSRA PPRLTVVSPN RATSTETSSS VETDLQPFRE AIALHGLSGA MASISVRSST PGSPTHVSSG SNASRRRNGL HDVDLNTFIS EEMALHLDNG GTRKRTSAQC GDVITDSPTH KRNRMI (1-275 aa encoded by B C113570)
C16orf72 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HUWE1-associated protein modifying stress responses 1; Telomere attrition and p53 response 1 protein
Gene Aliases: C16orf72; HAPSTR1; PRO0149; TAPR1
UniProt ID: (Human) Q14CZ0
Entrez Gene ID: (Human) 29035
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support