Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QELVGSEGA YKAAVDSFLQ QQHVLGAGGG PGPTQGEEQP PQPPLDPQNL ARPPAPAQTE ALSQLFCSVR TLTAKEELLQ TLRTHLILHM ARAHGYSKVM TGDSCTRLAI KLMTNLALGR GAFLAWDTGF SDERHGDVVV VRPMRDHTLK EVAFYNRLFS VPSVFTPAVD TKAPEKASIH RLMEAFILRL QTQFPSTVST VYRTSEKLVK GPRDGPAAGD SGPRCLLCMC ALDVDAADSA TAFGAQTSSR LSQMQSPIPL TETRTPPGPC CSPGVGWAQR CGQGACRRED PQACIEEQLC YSCRVNMKDL PSLDPLPPYI LAEAQLRTQR AWGLQEIRDC LIEDSDDEAG QS (165-515 aa encoded by BC125269)
Cytoplasmic tRNA 2-thiolation protein 2 (CUT2), also named as C16orf84 or NCS2, is a 515 amino acid protein, which belongs to the CUT2 family. C16orf84 localizes in the cytoplasm and Plays a central role in 2-thiolation of mcm5S2U at tRNA wobble positions of tRNA(Lys), tRNA(Glu) and tRNA(Gln). C16orf84 may act by forming a heterodimer with CTU1/ATPBD3 that ligates sulfur from thio-carboxylated URM1 onto the uridine of tRNAs at wobble position.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytoplasmic tRNA 2-thiolation protein 2; Cytosolic thiouridylase subunit 2; cytosolic thiouridylase subunit 2 homolog
Gene Aliases: 2310061F22Rik; C16orf84; CTU2; NCS2; UPF0432
UniProt ID: (Human) Q2VPK5, (Mouse) Q3U308
Entrez Gene ID: (Human) 348180, (Mouse) 66965
Molecular Function:
RNA metabolism protein
RNA processing factor
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support