Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 8
Immunogen sequence: ENQKILTPLV SLDTPGKATV QVVILADPDG HEICFVGDEA FRELSKMDPE GSKLLDDAMA ADKSDEWFAK HNKPKASG
Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 94%.
GLOD4 (Glyoxalase Domain Containing 4) is a Protein Coding gene. An important paralog of this gene is GLO1. [GeneCards]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glyoxalase domain-containing protein 4
Gene Aliases: 1700082G03Rik; 2700085E05Rik; C17orf25; C81254; CGI-150; GLOD4; HC71; My027; RGD1307010
UniProt ID: (Human) Q9HC38, (Mouse) Q9CPV4, (Rat) Q5I0D1
Entrez Gene ID: (Human) 51031, (Mouse) 67201, (Rat) 363644
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support