Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Component of chromatin complexes such as the MLL1/MLL and NURF complexes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BPTF associated protein of 18 kDa; BPTF-associated protein of 18 kDa; Chromatin complexes subunit BAP18; human embryo lung cellular protein interacting with SARS-CoV nsp-10; MLL1/MLL complex subunit C17orf49
Gene Aliases: BAP18; C17orf49; HEPIS
UniProt ID: (Human) Q8IXM2
Entrez Gene ID: (Human) 124944
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support