Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PETDLLAVAEH QALVFGGGPG RTSSEPPAGL RVSGEEETEN VGGANRHPRT SPKTSSCGVV HRPEREALEN EPGPQGTLS
Highest antigen sequence identity to the following orthologs - mouse 50%, rat 54%.
As part of the BORC complex may play a role in lysosomes movement and localization at the cell periphery. Associated with the cytosolic face of lysosomes, the BORC complex may recruit ARL8B and couple lysosomes to microtubule plus-end-directed kinesin motor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BLOC-1-related complex subunit 6; Lysosome-dispersing protein; Lyspersin
Gene Aliases: BORCS6; C17orf59; PRO2472
UniProt ID: (Human) Q96GS4
Entrez Gene ID: (Human) 54785
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support