Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
C17orf62 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytochrome b-245 chaperone 1; Eros; Essential for reactive oxygen species protein; HAPSTR1
Gene Aliases: C17orf62; CYBC1; EROS
UniProt ID: (Human) Q9BQA9
Entrez Gene ID: (Human) 79415
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support