Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IPTDQKVYQ SKPATLPRAK KMKGPIKDLI KAKGKELETE NEERNSKLVV DKPEQTVKTF PLPAVGLERA ATTKADKDIK NPIQPSFKML KNTKPMTTFQ EETKAQFYAS EKTSPKRELA KDLPKSGESR CNPSEAGASL LVGSIEPSLS NQDRKYSSTL PNDVQTTSGD LKLDKIDPQR QELLVKLLDV PTGDCHISPK NVSDGVKRVR TLLSNERDSK GRDHLSGV (35-261 aa encoded by BC005005 )
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell migration-inducing gene 3 protein; HLC-8; Human lung cancer oncogene 8 protein; lung cancer-related protein 8; Mitochondrial nucleoid-associated protein 1; Protein C17orf80; sperm-expressed protein 1
Gene Aliases: C17orf80; HLC-8; HLC8; MIG3; MTNAP1; SPEP1
UniProt ID: (Human) Q9BSJ5
Entrez Gene ID: (Human) 55028
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support