Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LSKDFCAREF EALRSCFAAA AKKTL
Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
NDUFAF8 is a protein coding gene involved in the assembly of mitochondrial NADH:ubiquinone oxidoreductase complex (complex I, MT-ND1). Required to stabilize NDUFAF5.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 8
Gene Aliases: C17orf89; NDUFAF8
UniProt ID: (Human) A1L188
Entrez Gene ID: (Human) 284184
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support