Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MRQKHYLEA AARGLHDSCP GQARYLLWAY TSSHDDKSTF EETCPYCFQL LVLDNSRVRL KPKARLTPKI QKLLNREARN YTLSFKEAKM VKKFKDSKSV LLITCKTCNR TVKHHGKSRS FVSTLKSNPA TPTSKLSLKT PERRTANPNH DMSGSKGKSP ASVFRTPTSG QSVSTCSSKN TSKTKKHFSQ LKMLLSQNES QKIPKVDFRN FLSSLKGGLL K (1-220 aa encoded by BC119766)
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HBV X-transactivated gene 13 protein; HBV X-transactivated protein 13; HBV XAg-transactivated protein 13; UPF0711 protein C18orf21
Gene Aliases: C18orf21; HsT3108; PNAS-124; PNAS-131; XTP13
UniProt ID: (Human) Q32NC0
Entrez Gene ID: (Human) 83608
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support