Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Protein kinase A (PKA)-binding protein. Binds to type II regulatory subunits of protein kinase A (PKA) and may block the A-kinase anchoring protein (AKAP)-mediated subcellular localization of PKA (PubMed:25653177).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A-kinase anchor protein C18orf42; A-kinase anchor protein inhibitor 1
Gene Aliases: AKAIN1; C18orf42
UniProt ID: (Human) P0CW23
Entrez Gene ID: (Human) 642597
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support