Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LLTKLSLIT APRRERGRPS LYVQRDIVQE TQREEDHRRE GLHVGRASTP DWVSEGPQPG LRRALSSDSI LSPAPDARAA DPAPEVRKVN RIPPDAYQPY LSPGTPQLEF SAFGAFGKPS SLSTAEAKAA TSPKATMSPR HLSESSGKPL STKQEASKPP RGCPQANRGV VRWEYFRLRP LRFRAPDEPQ QAQVPHVWGW EVAGAPALRL QKSQSSDLLE RERESVLRRE QEVAEERRNA LFPEVFSPTP DENSDQNSRS SSQASGITGS YSVSESPFFS PIHLHSNVAW TVEDPVDSAP PGQRKKEQWY AGINPSDGIN SEVLEAIRVT RHKNAMAERW ESRIYASEED D (330-679 aa encoded by BC052236)
The exact function of C19orf21 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: caprice; Mitotic interactor and substrate of PLK1; Mitotic spindle positioning protein
Gene Aliases: C19orf21; MISP; MISP1
UniProt ID: (Human) Q8IVT2
Entrez Gene ID: (Human) 126353
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support