Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Break repair meiotic recombinase recruitment factor 1; pre-T/NK cell associated protein (3B3); Pre-T/NK cell-associated protein 3B3
Gene Aliases: BRME1; C19orf57
UniProt ID: (Human) Q0VDD7
Entrez Gene ID: (Human) 79173
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support