This target displays homology in the following species: Dog: 77%; Human: 100%
C19orf67 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UPF0575 protein C19orf67; UPF0575 protein C19orf67-like
Gene Aliases: C19orf67
UniProt ID: (Human) A6NJJ6
Entrez Gene ID: (Human) 646457
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support