Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Human: 100%
C19orf68 gene ontology annotations related to this gene include multicellular organism development; ubiquitin-dependent protein catabolic process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Uncharacterized protein ZSWIM9
Gene Aliases: C19orf68; ZSWIM9
UniProt ID: (Human) Q86XI8
Entrez Gene ID: (Human) 374920
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support