Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Regulates drug efflux through modulation of ABCB1 localization and activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Required for drug-induced death protein 1
Gene Aliases: C1orf115; RDD1
UniProt ID: (Human) Q9H7X2
Entrez Gene ID: (Human) 79762
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support