Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTFQFNFTI EDHLENELTP IRDGALTLDS SKELSVSESQ KGEERDRKCS AEQFDLPQDH LWEHKSMENA APSQDTDSPL SAASSSRNLE PHGKQPSLRA AKEHAMPKDL KKMLENKVIE TLPGFQHVKL SVVKTILLKE NFPGENIVSK SFSSHSDLIT GVYEGGLKIW ECTFDLLAYF TKAKVKFAGK KVLDLGCGSG LLGITAFKGG SKEIHFQDYN SMVIDEVTLP NVVANSTLED EENDVNEPDV KRCRKPKVTQ LYKCRFFSGE WSEFCKLVLS SEKLFVKYDL ILTSETIYNP DYYSNLHQTF LRLLSKNGRV LLASKAHYFG VGGGVHLFQK FVEERDVFKT RILKIIDEGL KRFIIEITFK FPG (1-372 aa encoded by BC008679)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arsenic-transactivated protein 2; AsTP2; Histidine protein methyltransferase 1 homolog; Methyltransferase-like protein 18
Gene Aliases: ASTP2; C1orf156; HPM1; METTL18
UniProt ID: (Human) O95568
Entrez Gene ID: (Human) 92342
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support