Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arsenic-transactivated protein 2; AsTP2; Histidine protein methyltransferase 1 homolog; Methyltransferase-like protein 18
Gene Aliases: 2810422O20Rik; ASTP2; C1orf156; C78571; HPM1; METTL18; RGD1306783
UniProt ID: (Human) O95568, (Mouse) Q9CZ09, (Rat) Q4KM84
Entrez Gene ID: (Human) 92342, (Mouse) 69962, (Rat) 304928
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support