Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KEYGALVSEM KELRDLNRRL QDVLLLRLGS GPAIDLEKVK SECLEPEPEL RSTFSEEANT SSYYPAPAPV MDKYILDNGK VHLGSG
Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BEN domain-containing protein 5
Gene Aliases: BEND5; C1orf165
UniProt ID: (Human) Q7L4P6
Entrez Gene ID: (Human) 79656
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support